• USA Home
  • Search Results

  • dansylchloride2697560565211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dansyl chloride

3 Products
Synonym:   5-(Dimethylamino)naphthalene-1-sulfonyl chloride, DNSCl
Image of Dansyl chloride
CAS Number:
Empirical Formula (Hill Notation):
 C12H12ClNO2S
Molecular Weight:
 269.75
 Beilstein/REAXYS Number:
 2217205
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Suitability
  Product Line
  Solubility
Show More