• USA Home
  • Search Results

  • davephospdg376328144508587911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

DavePhos-Pd-G3

1 Products
Synonym:   Methanesulfonato 2-dicyclohexylphosphino-2-(N,N-dimethylamino)biphenyl(2′-amino-1,1′-biphenyl-2-yl) palladium(II)
Image of DavePhos-Pd-G3
CAS Number:
Empirical Formula (Hill Notation):
 C39H49N2O3PPdS
Molecular Weight:
 763.28
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Reaction Suitability
Show More

Product #
Description
804959 DavePhos-Pd-G3, 95%,