• USA Home
  • Search Results

  • dazomet1622853374411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dazomet

1 Products
Synonym:   3,5-Dimethyltetrahydro-1,3,5-thiadiazine-2-thione, Thiadiazine
Image of Dazomet
CAS Number:
Empirical Formula (Hill Notation):
 C5H10N2S2
Molecular Weight:
 162.28
 Beilstein/REAXYS Number:
 116039
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More