• USA Home
  • Search Results

  • dbhc2908331986811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

δ-BHC

1 Products
Synonym:   δ-1,2,3,4,5,6-Hexachlorocyclohexane solution, δ-HCH solution
CAS Number:
Empirical Formula (Hill Notation):
 C6H6Cl6
Molecular Weight:
 290.83
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More