• USA Home
  • Search Results

  • dcamphor1522346449311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

(+)-Camphor

6 Products
Synonym:   (1R)-(+)-Camphor, (1R)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one
Image of (+)-Camphor
CAS Number:
Empirical Formula (Hill Notation):
 C10H16O
Molecular Weight:
 152.23
 Beilstein/REAXYS Number:
 2042745
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Document Links
  Anal. Standard Form
  Shelf Life
  Allergen
  Organoleptic property
  Agency/Method
  Special Grade
  Physical Form
  Application(s)
  Assay
Show More