• USA Home
  • Search Results

  • ddecalactone1702570586211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

δ-Decalactone

2 Products
Synonym:   δ-Decanolactone, (±)-δ-Pentyl-δ-valerolactone, (±)-5-Decanolide, (±)-6-Pentyltetrahydro-2H-pyran-2-one, 5-Hydroxydecanoic acid δ-lactone
Image of δ-Decalactone
CAS Number:
Empirical Formula (Hill Notation):
 C10H18O2
Molecular Weight:
 170.25
 Beilstein/REAXYS Number:
 117520
 Council of Europe no.:
 621
 EC Number:
 FEMA Number:
 2361
 Flavis number:
 10.007
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More