• USA Home
  • Search Results

  • decaethyleneglycolmonododecylether62686900292011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Decaethylene glycol mono­dodecyl ether

1 Products
Synonym:   C12E10, Polyoxyethylene (10) lauryl ether
Image of Decaethylene glycol mono­dodecyl ether
CAS Number:
Empirical Formula (Hill Notation):
 C32H66O11
Molecular Weight:
 626.86
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More