• USA Home
  • Search Results

  • decamethoniumbromide4182954122011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Decamethonium bromide

1 Products
Synonym:   Decamethylene bis(trimethylammonium bromide), Decane-1,10-bis(trimethylammonium bromide)
Image of Decamethonium bromide
CAS Number:
Linear Formula:
 (CH3)3N(Br)(CH2)10N(Br)(CH3)3
Molecular Weight:
 418.29
 Beilstein/REAXYS Number:
 3728288
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  UV-Absorption Values
  Special Grade
  Assay
Show More