• USA Home
  • Search Results

  • decylbdmaltopyranoside482568249409511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Decyl β-D-maltopyranoside

1 Products
Synonym:   Decyl-β-D-maltoside
Image of Decyl β-<SC>D</SC>-maltopyranoside
CAS Number:
Empirical Formula (Hill Notation):
 C22H42O11
Molecular Weight:
 482.56
 Beilstein/REAXYS Number:
 4923334
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More