• USA Home
  • Search Results

  • deferoxaminemesylatesalt6567913814711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Deferoxamine mesylate salt

2 Products
Synonym:   DFOM, Deferoxamine methanesulfonate salt, Desferrioxamine mesylate salt
Image of Deferoxamine mesylate salt
CAS Number:
Empirical Formula (Hill Notation):
 C25H48N6O8 · CH4O3S
Molecular Weight:
 656.79
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
Show More