• USA Home
  • Search Results

  • deflazacort441521448447011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Deflazacort

2 Products
Synonym:   (11β,16β)-21-(acetyloxy)-11-hydroxy-2′-methyl-5′H-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione, 11β,21-dihydroxy-2′-methyl-5′βH-pregna-1,4-dieno[17,16-d]oxazole-3,20-dione 2′-acetate, DL-458-IT, L-5458, azacort, oxazacort, pregna-1,4-diene-11β,21-diol-3,20-dione[17α,16α-d]-2′-methyloxazoline 21-acetate
Image of Deflazacort
CAS Number:
Empirical Formula (Hill Notation):
 C25H31NO6
Molecular Weight:
 441.52
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More