• USA Home
  • Search Results

  • delphinidin3obdglucosidechloride50084690638311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Delphinidin 3-O-β-D-glucoside chloride

1 Products
Synonym:   Delphinidin 3-O-β-D-glucopyranoside chloride, Delphinidin 3-monoglucoside, Delphinin chloride, Myrtillin chloride
Image of Delphinidin 3-<I>O</I>-β-<SC>D</SC>-glucoside chloride
CAS Number:
Empirical Formula (Hill Notation):
 C21H21ClO12
Molecular Weight:
 500.84
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More