• USA Home
  • Search Results

  • delphinidinchloride3387052853011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Delphinidin chloride

3 Products
Synonym:   3,3′,4′,5,5′,7-Hexahydroxyflavylium chloride, 3,5,7-Trihydroxy-2-(3,4,5-trihydroxyphenyl)-1-benzopyrylium chloride
Image of Delphinidin chloride
CAS Number:
Empirical Formula (Hill Notation):
 C15H11ClO7
Molecular Weight:
 338.70
 EC Number:
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Anal. Standard Form
  Physical Form
  Special Grade
  Assay
Show More