• USA Home
  • Search Results

  • demeclocyclinehydrochloride501316473311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Demeclocycline hydrochloride

4 Products
Synonym:   7-Chloro-6-demethyltetracycline hydrochloride, Demethylchlorotetracycline hydrochloride
Image of Demeclocycline hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C21H21ClN2O8 · HCl
Molecular Weight:
 501.31
 Beilstein/REAXYS Number:
 3849198
 EC Number:
 MDL number:
 NACRES:
 NA.75
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Activity Spectrum
Show More