• USA Home
  • Search Results

  • densitystandard870kgm312345804247511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Density Standard 870 kg/m3

1 Products
Synonym:   Mineral oil
CAS Number:
 EC Number:
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More