• USA Home
  • Search Results

  • deoxycholicacid392578344311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Deoxycholic acid

4 Products
Synonym:   3α,12α-Dihydroxy-5β-cholanic acid, 7-Deoxycholic acid, Desoxycholic acid
Image of Deoxycholic acid
CAS Number:
Empirical Formula (Hill Notation):
 C24H40O4
Molecular Weight:
 392.57
 Beilstein/REAXYS Number:
 3219882
 EC Number:
 MDL number:
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Ice Shipping
  Functional group
  Physical Form
  Molecular weight
  ID clarifier
  Biological Source
Show More