• USA Home
  • Search Results

  • deoxynivalenol296325148110811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Deoxynivalenol

2 Products
Synonym:   3α,7α,15-Trihydroxy-12,13-epoxytrichothec-9-en-8-one, Vomitoxin
Image of Deoxynivalenol
CAS Number:
Empirical Formula (Hill Notation):
 C15H20O6
Molecular Weight:
 296.32
 MDL number:
 PubChem Substance ID:
Popular Documents:  Datasheet (PDF)  |  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Special Grade
Show More