• USA Home
  • Search Results

  • deoxynivalenolsolution296325148110811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Deoxynivalenol solution

2 Products
Synonym:   3α,7α,15-Trihydroxy-12,13-epoxytrichothec-9-en-8-one, DON, Vomitoxin
Image of Deoxynivalenol solution
CAS Number:
Empirical Formula (Hill Notation):
 C15H20O6
Molecular Weight:
 296.32
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Package quantity
  Anal. Standard Form
  Special Grade
  Concentration
Show More