• USA Home
  • Search Results

  • deoxyribonucleicacidfromcalfthymus123459108016911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Deoxyribonucleic acid from calf thymus

2 Products
Synonym:   Calf Thymus DNA, DNA from calf thymus, Thymonucleic acid
CAS Number:
 EC Number:
 MDL number:
 NACRES:
 NA.52
 eCl@ss:
 32160414
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Physical Form
Show More