• USA Home
  • Search Results

  • desipraminehydrochloride302845828611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Desipramine hydrochloride

5 Products
Image of Desipramine hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C18H22N2 · HCl
Molecular Weight:
 302.84
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Special Grade
  Anal. Standard Form
Show More