• USA Home
  • Search Results

  • desmethoxyyangonin228241534589811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Desmethoxyyangonin

2 Products
Synonym:   (E)-4-Methoxy-6-(2-phenylethenyl)-2H-pyran-2-one, 5,6-Dehydrokavain, Demethoxyyangonin
Image of Desmethoxyyangonin
CAS Number:
Empirical Formula (Hill Notation):
 C14H12O3
Molecular Weight:
 228.24
 Beilstein Registry Number:
 181238
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Titertest
Show More