• USA Home
  • Search Results

  • desoximetasone3764638267211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Desoximetasone

1 Products
Synonym:   9-Fluoro-11β,21-dihydroxy-16α-methylpregna-1,4-diene-3,20-dione
Image of Desoximetasone
CAS Number:
Empirical Formula (Hill Notation):
 C22H29FO4
Molecular Weight:
 376.46
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
Show More