• USA Home
  • Search Results

  • dexamethasone392465002211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dexamethasone

10 Products
Synonym:   (11β,16α)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16α-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione, 9α-Fluoro-16α-methylprednisolone, Prednisolone F
Image of Dexamethasone
CAS Number:
Empirical Formula (Hill Notation):
 C22H29FO5
Molecular Weight:
 392.46
 Beilstein/REAXYS Number:
 2066651
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Original Developer
  Potency
  Anal. Standard Form
  Special Grade
  Product Line
  Titertest
  Assay
  Physical Form
Show More