• USA Home
  • Search Results

  • dextromethorphanhydrobromide37032670034111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dextromethorphan hydrobromide

2 Products
Synonym:   (9S,13S,14S)-3-Methoxy-17-methylmorphinan hydrobromide, Dextromethorphan hydrobromide monohydrate
Image of Dextromethorphan hydrobromide
CAS Number:
Empirical Formula (Hill Notation):
 C18H25NO · HBr · H2O
Molecular Weight:
 370.32
 Beilstein Registry Number:
 6453793
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Anal. Standard Form
Show More