• USA Home
  • Search Results

  • dextrorphantartrate4074614398611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dextrorphan tartrate

2 Products
Synonym:   (+)-3-Hydroxy-N-methylmorphinan (+)-tartrate salt, Dextrorphan D-tartrate
Image of Dextrorphan tartrate
CAS Number:
Empirical Formula (Hill Notation):
 C17H23NO · C4H6O6
Molecular Weight:
 407.46
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More