• USA Home
  • Search Results

  • dextrose180165099711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dextrose

4 Products
Synonym:   D-(+)-Glucose, Dextrose
Image of Dextrose
CAS Number:
Empirical Formula (Hill Notation):
 C6H12O6
Molecular Weight:
 180.16
 Beilstein/REAXYS Number:
 1724615
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Ice Shipping
  Color
  Application(s)
  Agency/Method
Show More