• USA Home
  • Search Results

  • dfructose180165748711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D(-)Fructose

9 Products
Synonym:   D(-)Fructose, Laevulose, Levulose
Image of D(-)Fructose
CAS Number:
Empirical Formula (Hill Notation):
 C6H12O6
Molecular Weight:
 180.16
 EC Index Number:
 200-333-3
 MDL number
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More