• USA Home
  • Search Results

  • dgalactalcyclic34carbonate1721414984726711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-Galactal cyclic 3,4-carbonate

1 Products
Image of <SC>D</SC>-Galactal cyclic 3,4-carbonate
CAS Number:
Empirical Formula (Hill Notation):
 C7H8O5
Molecular Weight:
 172.14
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More