• USA Home
  • Search Results

  • dgalactose113c181157084930811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-Galactose-1-13C

2 Products
Image of <SC>D</SC>-Galactose-1-<SUP>13</SUP>C
CAS Number:
Empirical Formula (Hill Notation):
 13CC5H12O6
Molecular Weight:
 181.15
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Isotopic Purity
Show More