• USA Home
  • Search Results

  • dgluconicacidsolution1961652695411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-Gluconic acid solution

1 Products
Synonym:   2,3,4,5,6-Pentahydroxycaproic acid
Image of <SC>D</SC>-Gluconic acid solution
CAS Number:
Empirical Formula (Hill Notation):
 C6H12O7
Molecular Weight:
 196.16
 Beilstein/REAXYS Number:
 1726055
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
Show More