• USA Home
  • Search Results

  • dglucose2d181163073783811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-Glucose-2-d

1 Products
Synonym:   Deuterated Glucose, Dextrose-2-d, Labeled Glucose
Image of <SC>D</SC>-Glucose-2-d
CAS Number:
Linear Formula:
 HOCH2[CHOH]3CDOHCHO
Molecular Weight:
 181.16
 MDL number:
 PubChem Substance ID:
Popular Documents:  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More