• USA Home
  • Search Results

  • dglucuronicacid19414655612311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-Glucuronic acid

3 Products
Synonym:   Glucosiduronic acid
Image of <SC>D</SC>-Glucuronic acid
CAS Number:
Empirical Formula (Hill Notation):
 C6H10O7
Molecular Weight:
 194.14
 Beilstein/REAXYS Number:
 1727083
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Special Grade
Show More