• USA Home
  • Search Results

  • dhomoserine11912602721011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

D-Homoserine

1 Products
Synonym:   (R)-(+)-2-Amino-4-hydroxybutyric acid
Image of <SC>D</SC>-Homoserine
CAS Number:
Linear Formula:
 HOCH2CH2CH(NH2)CO2H
Molecular Weight:
 119.12
 Beilstein/REAXYS Number:
 1721680
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Physical Form
  Color
Show More

Product #
Description
H4021 D-Homoserine,