• USA Home
  • Search Results

  • diallylnndiisopropylphosphoramidite2453012642921811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diallyl N,N-diisopropylphosphoramidite

1 Products
Synonym:   N,N-Bis(1-methylethyl)phosphoramidous acid di-2-propenyl ester, Diallyloxy(diisopropylamino)phosphine
Image of Diallyl <I>N</I>,<I>N</I>-<WBR>diisopropylphosphoramidite
CAS Number:
Linear Formula:
 [(CH3)2CH]2NP(OCH2CH=CH2)2
Molecular Weight:
 245.30
 Beilstein Registry Number:
 4383514
 MDL number:
 PubChem Substance ID:
Popular Documents:  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More