• USA Home
  • Search Results

  • diallylphthalate2462613117911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diallyl phthalate

2 Products
Image of Diallyl phthalate
CAS Number:
Linear Formula:
 C6H4-1,2-(CO2CH2CH=CH2)2
Molecular Weight:
 246.26
 Beilstein/REAXYS Number:
 1880877
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More