• USA Home
  • Search Results

  • diaveridine26029535516811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diaveridine

3 Products
Synonym:   5-(3,4-Dimethoxybenzyl)-2,4-pyrimidinediamine
Image of Diaveridine
CAS Number:
Empirical Formula (Hill Notation):
 C13H16N4O2
Molecular Weight:
 260.29
 Beilstein/REAXYS Number:
 258464
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  Physical Form
  Special Grade
Show More