• USA Home
  • Search Results

  • diazepam2847443914511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diazepam

3 Products
Synonym:   7-Chloro-1-methyl-5-phenyl-3H-1,4-benzodiazepin-2(1H)-one, Ro 5-2807
Image of Diazepam
CAS Number:
Empirical Formula (Hill Notation):
 C16H13ClN2O
Molecular Weight:
 284.74
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Special Grade
  Manufacturer designation
Show More