• USA Home
  • Search Results

  • diazinondiethyld103144110015547311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diazinon-(diethyl-d10)

2 Products
Image of Diazinon-(<I>diethyl</I>-d<SUB>10</SUB>)
CAS Number:
Empirical Formula (Hill Notation):
 C12D10H11N2O3PS
Molecular Weight:
 314.41
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Isotopic Purity
Show More