• USA Home
  • Search Results

  • diazoxide2306736498711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Diazoxide

3 Products
Synonym:   7-Chloro-3-methyl-2H-1,2,4-benzothiadiazine 1,1-dioxide
Image of Diazoxide
CAS Number:
Empirical Formula (Hill Notation):
 C8H7ClN2O2S
Molecular Weight:
 230.67
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Product Information Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Special Grade
  Manufacturer designation
Show More