• USA Home
  • Search Results

  • dibenzo18crown6360401418732711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dibenzo-18-crown-6

1 Products
Synonym:   2,3,11,12-Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadeca-2,11-diene, 6,7,9,10,17,18,20,21-Octahydrodibenzo[b,k][1,4,7,10,13,16]hexaoxacyclooctadecene, DB18C6
Image of Dibenzo-18-crown-6
CAS Number:
Empirical Formula (Hill Notation):
 C20H24O6
Molecular Weight:
 360.40
 Beilstein/REAXYS Number:
 1162153
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
158399 Dibenzo-18-crown-6, 98%,