• USA Home
  • Search Results

  • dibenzothiophene1842613265011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dibenzothiophene

5 Products
Synonym:   DBT, Diphenylene sulfide
Image of Dibenzothiophene
CAS Number:
Empirical Formula (Hill Notation):
 C12H8S
Molecular Weight:
 184.26
 Beilstein/REAXYS Number:
 121101
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Anal. Standard Form
  Special Grade
  Assay
  Titertest
Show More