• USA Home
  • Search Results

  • dibenzylether1982610350411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dibenzyl ether

3 Products
Synonym:   Benzyl ether
Image of Dibenzyl ether
CAS Number:
Linear Formula:
 (C6H5CH2)2O
Molecular Weight:
 198.26
 Beilstein/REAXYS Number:
 1911156
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Application(s)
Show More