• USA Home
  • Search Results

  • dibutylether1302314296111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dibutyl ether

3 Products
Synonym:   Butyl ether
Image of Dibutyl ether
CAS Number:
Linear Formula:
 [CH3(CH2)3]2O
Molecular Weight:
 130.23
 Beilstein/REAXYS Number:
 1732752
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Purity Grade
  Potency
  Physical Form
  pH Value
  Solubility
Show More