• USA Home
  • Search Results

  • dibutylphthalate278348474211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dibutyl phthalate

7 Products
Synonym:   n-Butyl phthalate, DBP, Phthalic acid dibutyl ester
Image of Dibutyl phthalate
CAS Number:
Linear Formula:
 C6H4-1,2-[CO2(CH2)3CH3]2
Molecular Weight:
 278.34
 Beilstein/REAXYS Number:
 1914064
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Shelf Life
  Manufacturer designation
  Color
  Anal. Standard Form
  pH Value
  Product Line
  Application(s)
  Assay
  Physical Form
Show More
We're Sorry, no results were found.
Please try another search or browse our store using the categories on the left.