• USA Home
  • Search Results

  • dibutylsebacate3144610943311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dibutyl sebacate

7 Products
Synonym:   Decanedioic acid dibutyl ester, Sebacic acid dibutyl ester
Image of Dibutyl sebacate
CAS Number:
Linear Formula:
 [-(CH2)4CO2(CH2)3CH3]2
Molecular Weight:
 314.46
 Beilstein/REAXYS Number:
 1798308
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Functional group
  Refractive Index
  Solubility
  Application(s)
  Physical Form
  Assay
Show More