• USA Home
  • Search Results

  • dibutylsulfide1462954440111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dibutyl sulfide

5 Products
Synonym:   Butyl sulfide
Image of Dibutyl sulfide
CAS Number:
Linear Formula:
 CH3(CH2)3S(CH2)3CH3
Molecular Weight:
 146.29
 EC Index Number
 208-870-5
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Refractive Index
  Shelf Life
  Special Grade
Show More