• USA Home
  • Search Results

  • dibutyltindiacetate35103106733011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dibutyltin diacetate

2 Products
Synonym:   Diacetoxydibutyltin
Image of Dibutyltin diacetate
CAS Number:
Linear Formula:
 (CH3CH2CH2CH2)2Sn(OCOCH3)2
Molecular Weight:
 351.03
 Beilstein/REAXYS Number:
 4136035
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Potency
Show More