• USA Home
  • Search Results

  • dichloroaceticacid128947943611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dichloroacetic acid

3 Products
Image of Dichloroacetic acid
CAS Number:
Linear Formula:
 Cl2CHCOOH
Molecular Weight:
 128.94
 Beilstein/REAXYS Number:
 1098596
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Anal. Standard Form
  pH Value
  Vapor pressure
  Melting Point
  Assay
  Physical Form
  Special Grade
Show More