• USA Home
  • Search Results

  • dichloroacetonitrile10994301812011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dichloroacetonitrile

1 Products
Image of Dichloroacetonitrile
CAS Number:
Linear Formula:
 Cl2CHCN
Molecular Weight:
 109.94
 Beilstein/REAXYS Number:
 1739029
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
125954 Dichloroacetonitrile, 98%,