• USA Home
  • Search Results

  • dichlorodimethylsilane129067578511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Dichlorodimethylsilane

5 Products
Synonym:   Dimethyldichlorosilane
Image of Dichlorodimethylsilane
CAS Number:
Linear Formula:
 (CH3)2SiCl2
Molecular Weight:
 129.06
 EC Index Number
 014-003-00-X
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
Show More